Learn More
AdipoGen D-Biotin
SDP
Supplier:  AdipoGen CDXB0148G005
Encompass Procurement Services
Non-distribution item offered as a customer accommodation; additional freight charges may apply.
Learn More
Encompass_Preferred
Chemical. CAS 58-85-5. Formula C10H16N2O3S. MW 244.3. Synthetic. D--Biotin, also known as vitamin H or coenzyme R, is a water-soluble B-vitamin vitamin B7. D--Biotin is a cofactor responsible for carbon dioxide transfer in five carboxylase enzymes, such as acetyl-CoA carboxylase alpha, acetyl-CoA carboxylase beta, methylcrotonyl-CoA carboxylase, propionyl-CoA carboxylase or pyruvate carboxylase. It is important in fatty acid synthesis, branched-chain amino acid catabolism and gluconeogenesis. It covalently attaches to the epsilon-amino group of specific lysine residues in these carboxylases. This biotinylation reaction requires ATP and is catalyzed by holocarboxylase synthetase. Biotin is also covalently attached to distinct lysine residues in histones, affecting chromatin structure and mediating gene regulation. Biotin is widely used throughout the biotechnology industry to conjugate proteins for biochemical assays.
Catalog No. 50-256-1653
missing translation for 'QuantityLeft'