Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

D-Glucose, Anhydrous, Granular, ACS, Spectrum™ Chemical
SDP

Catalog No. p-7165449 Shop All Spectrum Chemical Manufacturing Corporation Products
Click to view available options
Quantity:
500 g
2.5 kg
12 kg
100 lb.
Packaging:
Fiber Drum
Poly Bottle
Poly Pail

Chemical Identifiers

CAS 50-99-7

Specifications

CAS Max % 1
Ignition Residue 0.0002
Loss on Drying 0.002
Packaging Fiber Drum
Molecular Formula C6H12O6
Quantity 100 lb.
Specific Rotation +52.5° to +53.0°
InChI Key WQZGKKKJIJFFOK-UHFFFAOYNA-N
SMILES OCC1OC(O)C(O)C(O)C1O
IUPAC Name 6-(hydroxymethyl)oxane-2,3,4,5-tetrol
Molecular Weight (g/mol) 180.16
Grade ACS
Insoluble Matter 5e-05
Titratable Acid 0.002 meq/g
Show More Show Less

CAUTION: For manufacturing or laboratory use only. Read and understand the label and Safety Data Sheet (SDS) prior to use.

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.