Learn More
Inmark Inc D-PAK FOR 6 VIALS
SDP
Supplier:  Inmark Inc 27233
Encompass Procurement Services
Non-distribution item offered as a customer accommodation; additional freight charges may apply.
Learn More
Encompass
This item has a minimum qty of 2 per supplier requirements.
Shipper; D-Pack; Frozen vials; EXAKT-PAK;; USDOT Regulations; IATA Packing Instruction 650 for diagnostic specimen transporting; 1 ft. EPS insulation
Catalog No. NC9199553