Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
d-Tartaric Acid, Crystal, BAKER ANALYZED™ A.C.S. Reagent, J.T.Baker™
Shop All Avantor Products
Click to view available options
Quantity:
500g
2.5kg
Packaging:
Poly Bottle
Specifications
Specifications
Formula Weight | 150.09 |
Physical Form | Crystals |
Grade | BAKER ANALYZED™ ACS Reagent |
Packaging | Poly Bottle |
Specific Gravity | 1L = 1.76kg |
Quantity | 500g |
Chemical Name or Material | d-Tartaric Acid |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction