Learn More
Eurofins Discoverx ChemiScreen D1 Dopamine Receptor Membrane Preparation, 1 vial. (HTS102M)
Supplier:  Eurofins Discoverx HTS102M
Encompass
The ChemiScreen D1 Dopamine Receptor Membrane Preparation is derived from proprietary stable recombinant cells to ensure high levels of DRD1 surface expression. ChemiScreen membrane preps can be used to screen for ligand binding affinities using a competitive radio-labeled ligand approach. Increased binding affinities will lead to higher radioactive signal. Membrane preps contain 1 mL of 1 mg/mL frozen membranes derived from a stably modified clonal cell line which was chosen for optimal receptor expression. For detailed product information, visit www.discoverx.com/membranepreps.
Catalog No. 50-200-5888
missing translation for 'QuantityLeft'
Explore all special offers
Add to Cart
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title
Your feedback has been submitted: Thank you for helping us improve our website.