Learn More
Eurofins Discoverx ChemiScreen D3 Dopamine Receptor Membrane Preparation, 1 vial. (HTS103M)
Supplier: Eurofins Discoverx HTS103M
The ChemiScreen D3 Dopamine Receptor Membrane Preparation is derived from proprietary stable recombinant cells to ensure high levels of DRD3 surface expression. ChemiScreen membrane preps can be used to screen for ligand binding affinities using a competitive radio-labeled ligand approach. Increased binding affinities will lead to higher radioactive signal. Membrane preps contain 1 mL of 1 mg/mL frozen membranes derived from a stably modified clonal cell line which was chosen for optimal receptor expression. For detailed product information, visit www.discoverx.com/membranepreps.
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.