Learn More
Medchemexpress LLC HY-15581S 1mg Medchemexpress, D8-MMAD CAS: Purity:>98%
SDP
Supplier:  Medchemexpress LLC HY15581S1MG
Encompass_Preferred
Medchemexpress, HY-15581S 1mg D8-MMAD CAS: D8-MMAD is a deuterated form of MMAD, which is a microtubule disrupting agent. Purity:>98% Medchemexpress has over 10000 novel life-science reagents, reference compounds, APIs and natural compounds for laboratory and scientific use. Other quantity can also be offered.
Catalog No. 50-193-2750
missing translation for 'QuantityLeft'
Explore all special offers
Add to Cart
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title
Your feedback has been submitted: Thank you for helping us improve our website.