Learn More
TARGETMOL CHEMICALS INC DAMGO TFA 78123-71-4FREE 5MG
Supplier:  TARGETMOL CHEMICALS INC T4411
Encompass Procurement Services
Non-distribution item offered as a customer accommodation; additional freight charges may apply.
Learn More
Encompass_Preferred
Also available in 1 mg 2 mg 10 mg 25 mg 50 mg and bulk. Please contact Fisher for quotes. Potent selective agonist of Mu-opioid receptor. Antinociceptive. Stimulates calcium-activated adenylyl cyclases related cAMP production. Expresses chemokines and chemokine receptors through TGF-beta1. Increases food intake of rats.
Catalog No. 50-257-9886