Learn More
Nasco DANi Teen and Young Adult Advanced Patient Care Simulator - Light

Shop All Simulaids, Inc. Products


DANi accurately reproduces scenarios and challenging dynamics for learners to administer critical care for this important patient demographic. Learners can practice on a comprehensive and complete simulation platform including patient monitoring capability and auscultation (heart and lung sounds).
A realistic, hands-on learning experience ideally suited for classroom, in-situ, or hospital instruction.
Basic Life Support (BLS) and Advanced Cardiac Life Support (ACLS)
CPR with quality performance metrics, carotid pulse, intubation
Trauma Simulation
- Physical assessment
- Airway, breathing, circulation
- Body trauma scan
- Wound care
- Self-harm/drug use moulage
- Asymmetric pupils
Realistic Features
- Pupils
- Airway
- Acne
- Smooth skin
- Tattoo sleeve
- Piercings and earrings
- Dyed and black wigs
- Foot wounds
Nursing Care
- Grooming
- Daily living assistance
- Eye/ear/hair care
- Catheter
- Injection
- Oxygen
- Blood pressure
- Pericare
- Ostomy care
- Urinary catheter care
- Oxygen administration
Female and Male Genitalia
- Includes female sexual assault genitalia
Patient Positioning
- Superior range of motion, seating, laying, finger and toe manipulation
Respiratory Support
- Manual ventilation with chest rise
- Intubation
- Ventilation
- Tracheostomy care
- Suctioning
Auscultation
- 6 anterior heart sites with 12 heart conditions
- 5 anterior, 6 upper posterior, 4 lower posterior
- 2 mid-axillary lung sites with 12 lung conditions
Advanced Skills
- Gastrostomy
- Nasogastric procedures
- Airway/trach suctioning
- Enema
- Pap smear
- Pelvic exam
- Intravenous injections
SKILLQUBE™ Patient Monitoring/Defibrillator
- ECG monitor (with additional patient vitals and monitored signals: ABP, SpO2, etCO2)
- Pacemaker
- Defibrillator
- AED
- Ventilator (generic)
- Instructor controlled scenario editor with library
- Auscultation, glucometer, thermometer, checklists, operating procedures
- Realistic laboratory values
- Optional CPR feedback with real-time evaluation (add-on)
Specifications
Product Type
Teen and Young Adult Patient Care Simulator
For Use With (Application)
Simulating Patient Care
Color
Light
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.