Learn More
Nasco DANi Teen and Young Adult Advanced Patient Skills Trainer - Medium

Shop All Simulaids, Inc. Products


DANi accurately reproduces scenarios and challenging dynamics for learners to administer critical care for this important patient demographic. Learners can also practice respiratory support, injection/IV with IM pads, intubation, CPR with quality performance metrics, non-invasive blood pressure, enema administration (female), catheterization, vaginal, pelvic and prostate examinations.
A realistic, hands-on learning experience ideally suited for classroom, in-situ, or hospital instruction.
Basic Life Support (BLS) and Advanced Cardiac Life Support (ACLS)
CPR with quality performance metrics, carotid pulse, intubation
Trauma Simulation
- Physical assessment
- Airway, breathing, circulation
- Body trauma scan
- Wound care
- Self-harm/drug use moulage
- Asymmetric pupils
Realistic Features
- Pupils
- Airway
- Acne
- Smooth skin
- Tattoo sleeve
- Piercings and earrings
- Dyed and black wigs
- Foot wounds
Nursing Care
- Grooming
- Daily living assistance
- Eye/ear/hair care
- Catheter
- Injection
- Oxygen
- Blood pressure
- Pericare
- Ostomy care
- Urinary catheter care
- Oxygen administration
Female and Male Genitalia
- Includes female sexual assault genitalia
Patient Positioning
- Superior range of motion, seating, laying, finger and toe manipulation
Respiratory Support
- Manual ventilation with chest rise
- Intubation
- Ventilation
- Tracheostomy care
- Suctioning
Advanced Skills
- Gastrostomy
- Nasogastric procedures
- Airway/trach suctioning
- Enema
- Pap smear
- Pelvic exam
- Intravenous injections
Specifications
Product Type
Teen and Young Adult Patient Skills Trainer
For Use With (Application)
Simulating Patient Care
Color
Medium
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.