Learn More
DAPT, Tocris Bioscience™
Shop All R&D Systems Products

Description
For more information about how DAPT may be used, see our protocols: Accelerated Induction of Cortical Neurons from hiPSCs, Human iPSC-derived Myoblasts, Generating Midbrain Dopaminergic Neurons from hPSCs .
DAPT synthesized to Ancillary Material Grade also available.
This product(s) resides on a Fisher Scientific GSA or VA contract. If you are viewing this page as a nonregistered user, the price(s) displayed is List Price. To view your GSA or VA contract pricing, log in using your account number, or become a registered user by contacting one of our Customer Service teams. You can also view your contract price by searching for this item(s) on GSA Advantage. To place an order, contact Fisher Scientific Customer Service.
Specifications
Specifications
Quantity | 10 mg |
Formula Weight | 432.46 |
Percent Purity | >99% |
Chemical Name or Material | DAPT |
Safety and Handling
Recommended Storage | Store at +4°C |
Your input is important to us. Please complete this form to provide feedback related to the content on this product.