Learn More
AdipoGen DAR-4M
SDP
Supplier:  AdipoGen CDXD0206M005
Encompass Procurement Services
Non-distribution item offered as a customer accommodation; additional freight charges may apply.
Learn More
Encompass_Preferred
Chemical. CAS 339527-79-6. Formula C25H26N4O3. MW 430.51. Synthetic Photo-stable fluorescent probe Ex ~560nm Em 575nm for the detection of nitric oxide NO in presence of oxygen, resulting in a triazolo-rhodamine analog DAR-4M T that exhibits about 840-fold greater fluorescence quantum efficiency. Suitable probe for monitoring RNS production, but not NO alone. Stable in a pH range of 4-12. Detection limit ~10nM.
Catalog No. 50-256-1183
missing translation for 'QuantityLeft'