Learn More
Medchemexpress LLC HY-12850A 50mg , Dasotraline (hydrochloride) SEP-225289 hydrochloride CAS:675126-08-6 Purity:98%
SDP
Supplier:  Medchemexpress LLC HY12850A50MG
Encompass_Preferred
Medchemexpress, HY-12850A 50mg Dasotraline (hydrochloride) SEP-225289 hydrochloride CAS:675126-08-6 Formula:C16H16Cl3N IC50: 4 nM (dopamine), 6 nM (norepinephrine), 11 nM (serotonin) Purity:98% Dasotraline hydrochloride (SEP-225289 hydrochloride) is a triple reuptake inhibitor that blocks dopamine , norepinephrine , and serotonin transporters with IC 50 values of 4, 6, and 11 nM, respectively. Medchemexpress has over 10000 novel life-science reagents, reference compounds, APIs and natural compounds for laboratory and scientific use. Other quantity can also be offered.
Catalog No. 50-149-3882
missing translation for 'QuantityLeft'
Explore all special offers
Add to Cart
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title
Your feedback has been submitted: Thank you for helping us improve our website.