Learn More
TARGETMOL CHEMICALS INC DAUNORUBICIN HYDROCHLORI 50MG
Supplier:  TARGETMOL CHEMICALS INC T1511
Encompass Procurement Services
Non-distribution item offered as a customer accommodation; additional freight charges may apply.
Learn More
Encompass_Preferred
Also available in 5 mg 10 mg 25 mg 100 mg 200 mg 500 mg 1 mL 10 mM (in DMSO) and bulk. Please contact Fisher for quotes. Daunorubicin hydrochloride (Rubidomycin hydrochloride) an anthracycline aminoglycoside antineoplastic inhibits DNA replication and repair and RNA and protein synthesis. purity: 99%
Catalog No. 50-257-7365