Learn More
Broadpharm NHS ester, 25 mg
Supplier:  Broadpharm BP22960
Encompass
DBCO-NHCO-PEG13-NHS ester is a long chain PEG linker containing NHS ester that is able to react specifically and efficiently with primary amines to form astable amide bond. The hydrophilic PEG13 arm improves water solubility and can also provide a long and flexible connection that minimizes steric hindrance during ligation. DBCO is commonly used for copper-free Click Chemistry reactions.
Catalog No. NC1763358
missing translation for 'QuantityLeft'
Explore all special offers
Add to Cart
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title
Your feedback has been submitted: Thank you for helping us improve our website.