Learn More
3M™ DBI-SALA™ ExoFit™ Construction Style Positioning Harness
Comfort fit with a single piece of material that wraps around you in the shape of an X. It dons without tangling and remembers your adjustments. Just snap three buckles. Shoulder, hip and leg padding can’t slip. A breathable lining guarantees you’ll stay dry and comfortable all day.
Supplier: 3M™ 1108502
Description
Specifications
Large | |
Back | |
6000 lb. | |
190 kg | |
Polyester |
n/a | |
Quick Connect | |
420 lb. | |
5.8 lb. |
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.