Learn More
Medchemexpress LLC HY-103371 5mg Medchemexpress, DCPIB CAS:82749-70-0 Purity:>98%
SDP
Supplier:  Medchemexpress LLC HY1033715MG
Encompass Procurement Services
Non-distribution item offered as a customer accommodation; additional freight charges may apply.
Learn More
Encompass_Preferred
Medchemexpress, HY-103371 5mg DCPIB CAS:82749-70-0 DCPIB is a selective, reversible and potent inhibitor of volume-regulated anion channels ( VRAC ), voltage-dependently activates potassium channels TREK1 and TRAAK, inhibits TRESK, TASK1 and TASK3 ( IC 50 s, 0.14, 0.95, 50.72 μM, respectively) [1] . DCPIB is also a selective blocker of swelling-induced chloride current ( I Cl,swell ), with an IC 50 of 4.1 μM in CPAE cells [2] . Purity:>98% Medchemexpress has over 10000 novel life-science reagents, reference compounds, APIs and natural compounds for laboratory and scientific use. Other quantity can also be offered.
Catalog No. 50-193-2242