Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Decafluorobenzophenone 98.0+%, TCI America™
SDP

Catalog No. p-7126836 Shop All Tokyo Chemical Industry (TCI) Products
Click to view available options
Quantity:
1 g
5 g

Chemical Identifiers

CAS 853-39-4
Molecular Formula C13F10O
Molecular Weight (g/mol) 362.126
MDL Number MFCD00000295
InChI Key WWQLXRAKBJVNCC-UHFFFAOYSA-N
Synonym decafluorobenzophenone, benzophenone, decafluoro, perfluorobenzophenone, methanone, bis pentafluorophenyl, bis perfluorophenyl methanone, bis pentafluorophenyl methanone, bis 2,3,4,5,6-pentafluorophenyl methanone, di2,3,4,5,6-pentafluorophenyl ketone, acmc-209q5n
PubChem CID 70068
IUPAC Name bis(2,3,4,5,6-pentafluorophenyl)methanone
SMILES C1(=C(C(=C(C(=C1F)F)F)F)F)C(=O)C2=C(C(=C(C(=C2F)F)F)F)F

Specifications

Melting Point 93°C
Color White-Yellow
Quantity 1 g
Formula Weight 362.13
Percent Purity ≥98.0% (GC)
Physical Form Crystalline Powder
Chemical Name or Material Decafluorobenzophenone
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.