Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Decylbenzene 98.0+%, TCI America™
SDP

Catalog No. p-7125995 Shop All Tokyo Chemical Industry (TCI) Products
Click to view available options
Quantity:
5 mL
25 mL

Chemical Identifiers

CAS 104-72-3
Molecular Formula C16H26
Molecular Weight (g/mol) 218.38
MDL Number MFCD00008955
InChI Key UZILCZKGXMQEQR-UHFFFAOYSA-N
Synonym 1-phenyldecane, benzene, decyl, n-decylbenzene, decane, 1-phenyl, n-decyl benzene, unii-kir29w0bp0, kir29w0bp0, phenyldecane, hsdb 920, decane, phenyl
PubChem CID 7716
IUPAC Name decylbenzene
SMILES CCCCCCCCCCC1=CC=CC=C1

Specifications

Melting Point -14°C
Color Colorless
Boiling Point 298°C
Quantity 5 mL
Formula Weight 218.38
Percent Purity ≥98.0% (GC)
Physical Form Liquid
Chemical Name or Material Decylbenzene
EINECSNumber (3)-0021
TSCA Yes
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.