Learn More
Bellco Glass, Inc. 3 Baff Nephelo Flask 300ML 14X130Mm
SDP
Supplier:  Bellco Glass, Inc. 257814133
Encompass_Preferred
Manufactured from type 1 class a Borosilicate glass. Ideal for growing, storing and mixing or aerobic and anaerobic cultures. The 3 bottom baffles increase culture oxygenation. Includes 25mm bellco stainless steel closure (2005 series) Sizes: 300ml
Catalog No. 50-194-5581
missing translation for 'QuantityLeft'
Explore all special offers
Add to Cart
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title
Your feedback has been submitted: Thank you for helping us improve our website.