Learn More
Hach Company Demineralizer Cartridge, UltraPure
Supplier: Hach Company 2079100
Color change indicator identifies when resin is exhausted. Largely free of silica and carbon dioxide. Designed for non-pressure applications, cartridge includes 3/8 inch barb hose connections on each end.
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.