Learn More
Perkin Elmer US LLC Demountable Platinum Injector, 1.5 mm I.D.
Supplier:  Perkin Elmer US LLC N8145108
Encompass_Preferred
This Platinum Injector Cassette Mounties designed for use with demountable torches in NexION 300/350 ICP-MS series instruments. The demountable design allows the injector to be removed and replaced efficiently without dismounting the torch. Standard Injectors have an O-ring free direct connection to Cyclonic and PFA spray chambers while O-ring free Injectors have an O-ring free seal that reduces contamination, makes it easy to clean and easy to insert.
Catalog No. 50-210-4854
missing translation for 'QuantityLeft'
Explore all special offers
Add to Cart
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title
Your feedback has been submitted: Thank you for helping us improve our website.