Learn More
A-M Systems Cold Curing Dental Cement Solvent, 16 oz can
SDP
Supplier:  A-M Systems 526000
Encompass Procurement Services
Non-distribution item offered as a customer accommodation; additional freight charges may apply.
Learn More
Encompass
This item has a minimum qty of 3 per supplier requirements.
16oz Cold Curing Dental Cement. This excellent bonding material can be used to affix stainless steel screws or cannula to exposed skull or bone, or as a general laboratory adhesive. Sets rapidly after mixing with the solvent.
Catalog No. NC9991372
missing translation for 'QuantityLeft'