Learn More
Electron Microscopy Sciences Dental Wax Pink 1 LB
Supplier: Electron Microscopy Sciences 72660
Pink wax is a hard and thick wax sheet that is ideal for mincing and cutting tissues. Red wax is no longer available. Approximately 31 to 35 sheets per pound.
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.