Learn More
Electron Microscopy Sciences Dental Wax Pink 5 LB
Supplier:  Electron Microscopy Sciences 72670
Encompass Procurement Services
Non-distribution item offered as a customer accommodation; additional freight charges may apply.
Learn More
Encompass_Preferred
Pink wax is a hard and thick wax sheet that is ideal for mincing and cutting tissues. Red wax is no longer available. Approximately 31 to 35 sheets per pound.
Catalog No. 50-949-027
missing translation for 'QuantityLeft'