Learn More
Quality Biological Inc Water, DEPC Treated, CS 10 X 1000 mL
Supplier: Quality Biological Inc 351068131CS
DEPC Treated Water is Ultra Pure and prepared by reverse osmosis, passed through fine carbon, de-ionized through two resin beds and serially filtered 1 ¿m positively charged membranes. ¿ The water is subsequently treated with 0.1% (v/v) DEPC. ¿ Used to reconstitute nucleic acids and/or proteins, and as a diluent or solvent for any molecular biology grade reagent. ¿ DEPC Treated Water may be usefully employed in RNA Applications (e.g., Preparations of an RNA probe, DNA Applications (e.g., Subcloning in pUC plasmids), and Protein Applications (e.g., Western Blotting). ¿ Storage Conditions: 15¿C to 30¿C ¿ Shipping Condition: ambient temperature ¿ Sterile Filtered & Deionized ¿ Endotoxin Tested < 0.03 EU/mL ¿ DNase, RNase, and Protease Tested ¿ Product and/or packaging contains recycled/renewable content ¿ Product and/or packaging is recyclable or reduces waste ¿ Product and/or packaging is safer to human and environmental health
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.