Learn More
Electron Microscopy Sciences DermaPlus Skin Protection for Dispenser 17 OZ
Supplier: Electron Microscopy Sciences 70571
A greaseless, non-toxic skin protector provides a barrier to aqueous-based materials and contains one of the most effective broad-spectrum antimicrobial agents available. It is formulated with lanolin, aloe vera, and vitamin E, and can be applied to any area of the skin.
DermaPlus is able to repel substances with any pH ranging from 0-14 (acids and bases except HF); caustic substances; and organic fluids. Prevents the continuation of irritation enabling the skin to heal naturally, even after repeated washings.
DermaPlus is not meant to be a glove replacement; however, it does offer added extra protection to the skin in case of glove failure or leakage. DermaPlus has been tested and is resistant to most chemicals used in microscopy. Wall mount dispenser required.
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.