Learn More
Electron Microscopy Sciences Desiccant Canister
Supplier:  Electron Microscopy Sciences 71210
Encompass_Preferred
Ideal for dry control of desiccator cabinets. Moisture is absorbed through the perforations in the aluminum case, which contains silica gel. This 2 x 4" canister maintains a safe level of humidity in 3cu.ft (0.8 cu. meter) area. When the monitor window turns pink, the silica gel is saturated. It can be regenerated in a 300 degree F (150 degree C) oven in about 3 hours. The monitor window will be blue in color when recharged.
Catalog No. 50-930-896
missing translation for 'QuantityLeft'
Explore all special offers
Add to Cart
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title
Your feedback has been submitted: Thank you for helping us improve our website.