Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Dexmedetomidine hydrochloride, Tocris Bioscience™
Shop All R&D Systems Products
Click to view available options
Quantity:
10 mg
50 mg
Description
Dexmedetomidine hydrochloride is an active isomer of medetomidine, a potent, highly selective α2-adrenoceptor agonist (Ki values are 1.08 and 1750 nM for α2- and α1-adrenoceptors respectively). Displays greater selectivity over α1-adrenoceptors than Clonidine (Cat. No. 0690) and UK 14,304 (1620-, 220- and 300-fold respectively). Active in vivo; displays hypotensive, bradycardic, sedative, anxiolytic, hypothermic and analgesic effects.
Racemate also available.
Specifications
Specifications
Quantity | 10 mg |
Formula Weight | 236.74 |
Percent Purity | 98% |
Chemical Name or Material | Dexmedetomidine hydrochloride |
Safety and Handling
Recommended Storage | Desiccate at Room Temperature |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction