Learn More
DHX57 Polyclonal Antibody, Bethyl Laboratories

Rabbit Polyclonal Antibody
$154.46
Specifications
Antigen | DHX57 |
---|---|
Applications | Western Blot |
Classification | Polyclonal |
Conjugate | Unconjugated |
Form | Liquid |
Catalog Number | Mfr. No. | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Catalog Number | Mfr. No. | Quantity | Price | Quantity & Availability | |||||
50-156-2255
![]() |
Bethyl Laboratories, Inc
A302300AT |
20 μL |
Each of 1 for $154.46
|
|
|||||
Description
The recommended shelf life for this product is 1 year from date of receipt.
Probable ATP-binding RNA helicase.Specifications
DHX57 | |
Polyclonal | |
Liquid | |
RUO | |
TBS with 0.1% BSA and 0.09% sodium azide | |
DDX57, DEAH box protein 57, DEAH-box RNA/DNA helicase AAM73547, putative ATP-dependent RNA helicase DHX57 | |
DHX57 | |
IgG | |
Antigen affinity chromatography | |
Antibody |
Western Blot | |
Unconjugated | |
Rabbit | |
Human | |
Q6P158 | |
90957 | |
Between 150 and 200 | |
Primary | |
4° C | |
DHX57 |
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.