Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Diallyl trisulfide, 98.8%, MP Biomedicals™

Catalog No. p-4405622 Shop All MP Biomedicals, Inc Products
Click to view available options
Quantity:
25 mg
0.1 g
0.5 g

Chemical Identifiers

CAS 2050-87-5
Molecular Formula C6H10S3
Molecular Weight (g/mol) 178.326
InChI Key UBAXRAHSPKWNCX-UHFFFAOYSA-N
Synonym diallyl trisulfide, allyl trisulfide, allitridin, trisulfide, di-2-propenyl, diallyltrisulfide, diallyl trisulphide, allitridum, dasuansu, diallyltrisulfane, di-2-propenyl trisulfide
PubChem CID 16315
ChEBI CHEBI:78492
IUPAC Name 3-(prop-2-enyltrisulfanyl)prop-1-ene
SMILES C=CCSSSCC=C

Specifications

Color Colorless
Assay Percent Range 0.988
Quantity 25 mg
Formula Weight 178.3
Physical Form Liquid
Chemical Name or Material DATS
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.