Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Diammonium Hydrogen Citrate, 15% (w/w), Reagents
Click to view available options
:
Quantity: 1 L
Specifications
Specifications
Color | Colorless |
Boiling Point | 100°C |
Packaging | Poly Bottle |
Quantity | 1 L |
Specific Gravity | 1.052 |
Percent Purity | 15% |
Grade | Reagent |
Physical Form | Liquid |
Chemical Name or Material | Diammonium Hydrogen Citrate, 15% (w/w) |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction