Learn More
Electron Microscopy Sciences Diamond Disc 10" 120 grit, PSA backing
Supplier: Electron Microscopy Sciences 5027008
Flexible Diamond Discs will reduce the number of steps in the grinding/polishing process along with a decrease in preparation time. Our Flexible Diamond Discs replace traditional SiC discs for most applications and give a superior finish and perfectly flat specimens. The patterned design minimizes build up of abraded material.
Particles: 120 grit
Backing: PSA
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.