Learn More
Electron Microscopy Sciences Diamond Disc 12" 80 grit, PSA Steel Plate Back
Supplier: Electron Microscopy Sciences 5028006
Flexible Diamond Discs will reduce the number of steps in the grinding/polishing process along with a decrease in preparation time. Our Flexible Diamond Discs replace traditional SiC discs for most applications and give a superior finish and perfectly flat specimens. The patterned design minimizes build up of abraded material.
Particles: 80 grit
Backing: PSA Steel Plate
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.