Learn More
Electron Microscopy Sciences Diamond Finishing Disc, Extra Fine Grit
Supplier:  Electron Microscopy Sciences 6214625
Encompass_Preferred
Diamond Finishing discs offer extremely aggressive grinding action, yet leave an exceptionally smooth finish. Discs have pressure sensitive adhesive backing for a secure fit on holder and 3/4" (19.1 mm) diameter.
Grit: Extra Fine
Color: Blue
Catalog No. 50-996-842
missing translation for 'QuantityLeft'
Explore all special offers
Add to Cart
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title
Your feedback has been submitted: Thank you for helping us improve our website.