Learn More
Anton Paar 1kg package of Diatomaceous Earth
Supplier:  Anton Paar 164788
Encompass Procurement Services
Non-distribution item offered as a customer accommodation; additional freight charges may apply.
Learn More
Encompass_Preferred
Diatomaceous Earth with the highest clarification efficiency used for enhanced sample preparation of turbid samples of beer and wine. Refill for item Alex 500 sample preparation kit item NC1818625
Catalog No. NC1818626
missing translation for 'QuantityLeft'