Learn More
Medchemexpress LLC HY-B0926 500mg Medchemexpress, Diatrizoic acid CAS:117-96-4 Purity:>98%
SDP
Supplier:  Medchemexpress LLC HYB0926500MG
Encompass Procurement Services
Non-distribution item offered as a customer accommodation; additional freight charges may apply.
Learn More
Encompass_Preferred
Medchemexpress, HY-B0926 500mg Diatrizoic acid CAS:117-96-4 Diatrizoic acid (Diatrizoate) is an iodinated radiocontrast agent and has the potential for radiographic imaging of the airways. Diatrizoic acid induces mitochondrial turnover and oxidative stress, and activating apoptosis by dysregulating calcium. Purity:>98% Medchemexpress has over 10000 novel life-science reagents, reference compounds, APIs and natural compounds for laboratory and scientific use. Other quantity can also be offered.
Catalog No. 50-208-2671