Learn More
Medchemexpress LLC HY-Y0304 50mg Medchemexpress, Dibutyl phthalate CAS:84-74-2 Purity:>98%
Supplier: Medchemexpress LLC HYY030450MG
Medchemexpress, HY-Y0304 50mg Dibutyl phthalate CAS:84-74-2 Dibutyl phthalate is a commonly used plasticizer commonly found in some food packaging materials, personal care products, and the coating of oral medications [1] . May cause toxicity and adverse neurobehavioral effects [2] [3] . Purity:>98% Medchemexpress has over 10000 novel life-science reagents, reference compounds, APIs and natural compounds for laboratory and scientific use. Other quantity can also be offered.
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.