Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Dichloramine B 95.0+%, TCI America™
SDP

Catalog No. p-7126139 Shop All Tokyo Chemical Industry (TCI) Products
Click to view available options
Quantity:
5 g
25 g

Chemical Identifiers

CAS 473-29-0
Molecular Formula C6H5Cl2NO2S
Molecular Weight (g/mol) 226.07
MDL Number MFCD00025025
InChI Key PJBJJXCZRAHMCK-UHFFFAOYSA-N
Synonym Benzenesulfonedichloramide
PubChem CID 68055
IUPAC Name N,N-dichlorobenzenesulfonamide
SMILES ClN(Cl)S(=O)(=O)C1=CC=CC=C1

Specifications

Melting Point 74°C
Quantity 5 g
UN Number 1479
Formula Weight 226.07
Percent Purity ≥95.0% (T)
Physical Form Crystalline Powder
Chemical Name or Material Dichloramine B
EINECSNumber (3)-3131
TSCA Yes
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.