Learn More
Dichloro(diethylamino)phosphine, 97%, Thermo Scientific Chemicals
Description
Dichloro(diethylamino)phosphine is used as a reagent for the preparation of diethylaminophosphine-metal complexes and it is a critical starting material in the preparation of various phosphines, phosphoramidites & the phosphitylation of alcohols.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Dichloro(diethylamino)phosphine is used as a reagent for the preparation of diethylaminophosphine-metal complexes and it is a critical starting material in the preparation of various phosphines, phosphoramidites & the phosphitylation of alcohols.
Solubility
Reacts violently in water.
Notes
Moisture sensitive. Store away from oxidizing agents, bases, active metals, water/ moisture.
Specifications
Specifications
Density | 1.196 |
Boiling Point | 179°C |
Flash Point | 83°C (181°F) |
Odor | Irritating |
Refractive Index | 1.497 |
Linear Formula | (CH3CH2)2NPCl2 |
Quantity | 5 g |
UN Number | UN3265 |
Sensitivity | Moisture sensitive |
Solubility Information | Reacts violently in water. |
Show More |
Safety and Handling
Hazard Category | H227-H314 |
Hazard Statement | GHS H Statement H314-H318-H227 Causes severe skin burns and eye damage. Causes serious eye damage. Combustible liquid. |
Precautionary Statement | P210-P260-P264b-P280-P301+P330+P331-P303+P361+P353-P304+P340-P305+P351+P338-P310-P363-P370+P378q-P501c |
DOTInformation | Transport Hazard Class: 8; Packing Group: II; Proper Shipping Name: CORROSIVE LIQUID, ACIDIC, ORGANIC, N.O.S. |
TSCA | No |
Recommended Storage | Ambient temperatures |
RUO – Research Use Only
Your input is important to us. Please complete this form to provide feedback related to the content on this product.