Learn More
TARGETMOL CHEMICALS INC DICHLORPHENAMIDE 100MG
Also available in 5 mg 10 mg 25 mg 50 mg 200 mg 500 mg 1 mL 10 mM (in DMSO) and bulk. Please contact Fisher for quotes. Dichlorphenamide (Diclofenamide) is a carbonic anhydrase inhibitor that is used in the treatment of glaucoma. purity: 98%