Learn More
Selleck Chemical LLC Dichlorphenamide 10mg 120-97-8 Diclofenamide
Supplier:  Selleck Chemical LLC S217710MG
Encompass_Preferred
Dichlorphenamide (Diclofenamide) is a sulfonamide and a carbonic anhydrase inhibitor of the meta-Disulfamoylbenzene class.
Catalog No. 50-524-9
missing translation for 'QuantityLeft'
Add to Cart
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title
Your feedback has been submitted: Thank you for helping us improve our website.