Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Didecyl Phosphate 95.0+%, TCI America™
SDP

Catalog No. p-7134899 Shop All Tokyo Chemical Industry (TCI) Products
Click to view available options
Quantity:
100 mg
1 g
5 g

Chemical Identifiers

CAS 7795-87-1
Molecular Formula C20H43O4P
Molecular Weight (g/mol) 378.53
MDL Number MFCD00015062
InChI Key QHAUASBJFFBWMY-UHFFFAOYSA-N
Synonym Phosphoric Acid Didecyl Ester
PubChem CID 82260
IUPAC Name bis(decyloxy)phosphinic acid
SMILES CCCCCCCCCCOP(O)(=O)OCCCCCCCCCC

Specifications

Melting Point 47°C
Color White
Quantity 100 mg
Formula Weight 378.53
Percent Purity ≥95.0% (T)
Physical Form Crystalline Powder
Chemical Name or Material Didecyl Phosphate
EINECSNumber (2)-1986
TSCA Yes
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.