Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Diethyltin dichloride, Thermo Scientific Chemicals

Catalog No. p-7080325
Click to view available options
Quantity:
0.5 g
2 g
10 g

Used as organic intermediates and active pharmaceutical intermediate.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Chemical Identifiers

CAS 866-55-7
Molecular Formula C4H10Cl2Sn
Molecular Weight (g/mol) 247.734
MDL Number MFCD00013609
InChI Key YFAXVVMIXZAKSR-UHFFFAOYSA-L
Synonym diethyltin dichloride, diethyldichlorotin, diethyltindichloride, dichlorodiethyltin, diethyldichlorostannane, tin, dichlorodiethyl, stannane, dichlorodiethyl, diethyltin chloride, tin, diethyl-, dichloride, unii-pga900d045
PubChem CID 91491
IUPAC Name dichloro(diethyl)stannane
SMILES CC[Sn](CC)(Cl)Cl

Specifications

Melting Point 84°C
Color White
Boiling Point 220°C
Linear Formula (C2H5)2SnCl2
Quantity 0.5 g
Sensitivity Moisture sensitive
Solubility Information Partly soluble in water((0.023g/L) (25°C),Calc).
Formula Weight 247.72
Physical Form Crystalline
Chemical Name or Material Diethyltin dichloride
Hazard Category H302-H315-H319-H335
Hazard Statement GHS H Statement
H302-H315-H319-H335
Harmful if swallowed.
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Precautionary Statement P261-P264b-P270-P271-P280-P301+P312-P302+P352-P304+P340-P305+P351+P338-P312-P330-P332+P313-P362-P501c
EINECSNumber 212-750-8
RTECSNumber WH7200000
TSCA No
Recommended Storage Ambient temperatures

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.