Learn More
Fisher Science Education™ Digital Dry Bath Block
SCI_ED
Catalog No. S29031
Click to view available options
No. of Places:
Precision machined to match the conical size and shape of 0.5, 1.5, 2.0, 15 and 50mL tubes. These high-grade aluminum blocks are unsurpassed in temperature uniformity.
These high-grade aluminum blocks are precision machined to match the conical size and shape of 0.5, 1.5, 2.0, 15 and 50mL tubes - providing unsurpassed temperature uniformity.
Order Info
Blocks are sold separately from Dry Bath; see S29028 or S29029 for associated Dry Bath.
Certifications
CE
Specifications
For Use With (Equipment)
Digital Dry Baths
Material
Aluminum
No. of Places
24
Type
Block
For Use With (Application)
Heating and incubating samples
Tube Size
1.5 ml or 2.0mL
Warranty
2 Years
Dimensions (L x W x H)
7.62 x 9.53 x 5.08 cm (3 x 3.75 x 2 in.)