Learn More
Fisher Science Education™ Digital Dry Bath Block
SCI_ED
Catalog No. S29039
Click to view available options
Holds:
Precision machined to match the conical size and shape of 0.5, 1.5, 2.0, 15 and 50mL tubes. These high-grade aluminum blocks are unsurpassed in temperature uniformity.
These high-grade aluminum blocks are precision machined to match the conical size and shape of 0.5, 1.5, 2.0, 15 and 50mL tubes - providing unsurpassed temperature uniformity.
Order Info
Blocks are sold separately from Dry Bath; see S29028 or S29029 for associated Dry Bath.
Certifications
CE
Specifications
For Use With (Equipment)
Two and Four-Block Digital Dry Baths
Holds
96 well PCR Plate
Material
Aluminum
No. of Places
1
Type
Block
For Use With (Application)
Heating and incubating samples
Tube Size
0.2mL
Warranty
2 Years
Dimensions (L x W x H)
7.62 x 9.53 x 5.08 cm (3 x 3.75 x 2 in.)