Learn More
Stoelting™ Digital Manipulator Arm
SDP
Digital Stereotaxic Manipulator Arms have sealed electronic sensors, highly immune to outside interference and environmental conditions on each axis.
$5570.00 - $7825.00
Specifications
For Use With (Application) Neuroscience
Size 3 axes
View More Specs
Products 4
Catalog Number Mfr. No. Model Price Quantity & Availability  
Catalog Number Mfr. No. Model Price Quantity & Availability  
10-000-232
SDP
View Documents
Stoelting™
51904
Add-on, Left
Each for $7,818.00
missing translation for 'QuantityLeft'
Add to Cart
10-000-233
SDP
View Documents
Stoelting™
51906
Add-on, Right
Each for $7,825.00
missing translation for 'QuantityLeft'
Add to Cart
10-000-234
SDP
View Documents
Stoelting
51914
Conversion, Left
Each for $5,570.00
missing translation for 'QuantityLeft'
Add to Cart
10-000-235
SDP
View Documents
Stoelting
51916
Conversion, Right
Each for $5,570.00
missing translation for 'QuantityLeft'
Add to Cart
Description
Description
  • Measurements are accurate to 10 microns in all three directions
  • Movements are monitored by a large, easy-to-read LED display module
  • Resolution as precise as 10 microns
  • Digital stereotaxic arm allows for:
    • Memory of target coordinates
    • Zeroing function at reference point
    • 10-micron digital scale
  • Upgrading an existing stereotaxic to a digital model will reduce the risk of human error and increase accuracy (10-rather than 100-micron resolution)
  • Large LED display is also useful for surgeries performed in low light, or constrained environments, where reading the vernier scale is difficult
Options:
  • Add-on Motorized Manipulator Arm and Axis StereoDrive Software
    Keep existing arm and have a new one shipped
  • Conversion to Motorized Manipulator Arm and Axis StereoDrive Software
    Send existing arm for conversion to motorized
Specifications
Specifications
Neuroscience
3 axes
Videos
SDS
Documents
Documents
Product Certifications
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Your feedback has been submitted: Thank you for helping us improve our website.