Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Digoxin 96.0+%, TCI America™
Shop All Tokyo Chemical Industry (TCI) Products

Click to view available options
Quantity:
100 mg
1 g
Specifications
Specifications
Melting Point | 265°C |
Color | White-Yellow |
Quantity | 100 mg |
UN Number | 1544 |
Formula Weight | 780.95 |
Percent Purity | ≥96.0% (HPLC) |
Physical Form | Crystalline Powder |
Chemical Name or Material | Digoxin |
Safety and Handling
RTECSNumber | IH6125000 |
TSCA | Yes |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction