Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Dihydro-β-erythroidine hydrobromide, Tocris Bioscience™
GSA_VA

Catalog No. p-4783907 Shop All R&D Systems Products
Click to view available options
Quantity:
10 mg
50 mg
This item is not returnable. View return policy
This item is not returnable. View return policy

α4β2, muscle type and Torpedo nAChR antagonist

Dihydro-β-erythroidine hydrobromide is a member of the Erythrina alkaloids, it is a competitive nicotinic acetylcholine receptor antagonist with moderate selectivity for the neuronal α4 receptor subunit (IC50 values are 0.19 and 0.37 μM for α4β4 and α4β2 receptors, respectively). Antagonizes behavioral effects of nicotine in vivo. Dihydro-β-erythroidine blocks excitation of striatal GABAergic neurons, which completely suppress polysynaptic inhibition between striatal cholinergic interneurons. Dihydro-β-erythroidine hydrobromide has antidepressive-like effects in mice (forced swim and mouse suspension). Orally bioavailable

This product(s) resides on a Fisher Scientific GSA or VA contract. If you are viewing this page as a nonregistered user, the price(s) displayed is List Price. To view your GSA or VA contract pricing, log in using your account number, or become a registered user by contacting one of our Customer Service teams. You can also view your contract price by searching for this item(s) on GSA Advantage. To place an order, contact Fisher Scientific Customer Service.

Chemical Identifiers

CAS 29734-68-7
Molecular Formula C16H22BrNO3
Molecular Weight (g/mol) 356.26
InChI Key GFIGWAJEIMHJJB-LINSIKMZSA-N
Synonym dihydro-beta-erythroidine hydrobromide, unii-ns1u03gc05, 2s,13bs-2-methoxy-2,3,5,6,8,9,10,13-octahydro-1h,12h-benzo i pyrano 3,4-g indolizin-12-one hydrobromide, dh?e, dihydro-?-erythroidine hydrobromide, 3, a-1,6-didehydro-14,17-dihydro-3-methoxy-16 15h-oxaerythrinan-15-one hydrobromide, 3beta-1,6-didehydro-14,17-dihydro-3-methoxy-16 15h-oxaerythrinan-15-one hydrobromide
PubChem CID 11957537
SMILES COC1CC=C2CCN3C2(C1)C4=C(CC3)COC(=O)C4.Br

Specifications

Quantity 10 mg
Formula Weight 356.26
Percent Purity >98%
Chemical Name or Material Dihydro-β-erythroidine hydrobromide
Recommended Storage Desiccate at Room Temperature
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.