Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Diisopropyl Adipate, +99%, Spectrum™ Chemical
SDP

Catalog No. p-7226121 Shop All Spectrum Chemical Manufacturing Corporation Products
Click to view available options
Quantity:
25 mL
500 mL

Synonym: Adipic Acid Diisopropyl Ester, Isopropyl Adipate, Dipropan-2-yl Hexanedioate

  • Used in cosmetic and personal care products as an emollient and fragrance
  • Ungraded products are indicative of a grade suitable for general industrial use or research purposes and typically are not suitable for human consumption or therapeutic use

Chemical Identifiers

CAS 6938-94-9

Specifications

Quantity 25 mL
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.